ENA321762637 Enamine

1-(2-chloropropanoyl)-3-prop-2-en-1-ylurea

  • MDL number MFCD11185278
  • Cas Number : 1094302-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29336

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29336
Availability details for
Quantity
  • Availability
  • Add to Cart