ENA321762643 Enamine

4-(2-methyl-1,3-oxazol-4-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD09864967
  • Cas Number : 1086376-81-9
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-methyl-1,3-oxazol-4-yl)benzenesulfonyl chloride
Inchi Code 1S/C10H8ClNO3S/c1-7-12-10(6-15-7)8-2-4-9(5-3-8)16(11,13)14/h2-6H,1H3
Inchi Key HPSZYMHIXLWICC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74193

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-74193
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart