ENA321762647 Enamine

2-[(3-aminopiperidin-1-yl)methyl]-4-chlorophenol

  • MDL number MFCD14617598
  • Cas Number : 1249550-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74197
Availability details for
Quantity
  • Availability
  • Add to Cart