ENA321762649 Enamine

2-amino-5-(1,3-benzoxazol-2-ylsulfanyl)-2-methylpentanenitrile

  • MDL number MFCD15452625
  • Cas Number : 1308025-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74199

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74199
Availability details for
Quantity
  • Availability
  • Add to Cart