ENA321762651 Enamine

2-(3-chlorophenyl)-2-(methylsulfanyl)ethan-1-amine

  • MDL number MFCD16062547
  • Cas Number : 1248601-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74201
Availability details for
Quantity
  • Availability
  • Add to Cart