ENA321762652 Enamine

tert-butyl N-(2-amino-4-methylpentyl)carbamate

  • MDL number MFCD14584431
  • Cas Number : 1117693-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74203

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74203
Availability details for
Quantity
  • Availability
  • Add to Cart