ENA321762653 Enamine

1-phenyl-4-(piperidin-3-yl)piperazine dihydrochloride

  • MDL number MFCD18483269
  • Cas Number : 1281095-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 296...298
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74205

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74205
Availability details for
Quantity
  • Availability
  • Add to Cart