ENA321762655 Enamine

2-(2,2,2-trifluoroethoxy)ethane-1-sulfonyl chloride

  • MDL number MFCD14678417
  • Cas Number : 212190-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74210
Availability details for
Quantity
  • Availability
  • Add to Cart