ENA321762659 Enamine

1-(furan-2-yl)-2-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD16661006
  • Cas Number : 1306604-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74215
Availability details for
Quantity
  • Availability
  • Add to Cart