ENA321762663 Enamine

N-cyclohexyl-2-hydroxyacetamide

  • MDL number MFCD08442132
  • Cas Number : 90204-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-66
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74220
Availability details for
Quantity
  • Availability
  • Add to Cart