ENA321762669 Enamine

2-methyl-3-(phenylsulfanyl)propanoic acid

  • MDL number MFCD12132651
  • Cas Number : 775-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40...42
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74229

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74229
Availability details for
Quantity
  • Availability
  • Add to Cart