ENA321762671 Enamine

3,6-dichloro-1-benzothiophene-2-carboxamide

  • MDL number MFCD11643423
  • Cas Number : 34586-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269...271
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74232

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74232
Availability details for
Quantity
  • Availability
  • Add to Cart