ENA321762673 Enamine

2-(1H-pyrrol-2-yl)ethan-1-amine

  • MDL number MFCD08448797
  • Cas Number : 40808-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54456
Availability details for
Quantity
  • Availability
  • Add to Cart