ENA321762674 Enamine

4-(4-methylphenyl)oxane-4-carboxylic acid

  • MDL number MFCD08445816
  • Cas Number : 889940-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74237

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74237
Availability details for
Quantity
  • Availability
  • Add to Cart