ENA321762681 Enamine

3-chloro-6-cyclopropylpyridazine

  • MDL number MFCD14702638
  • Cas Number : 1046816-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74245
Availability details for
Quantity
  • Availability
  • Add to Cart