ENA321762684 Enamine

2-(phenylamino)-1-(piperidin-1-yl)ethan-1-one

  • MDL number MFCD01700292
  • Cas Number : 92032-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74248
Availability details for
Quantity
  • Availability
  • Add to Cart