ENA321762691 Enamine

1-(2-chloropropanoyl)-3-(2,2,2-trifluoroethyl)urea

  • MDL number MFCD11103772
  • Cas Number : 1152566-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29358
Availability details for
Quantity
  • Availability
  • Add to Cart