ENA321762693 Enamine

2-[(4-methylphenyl)amino]-1-(piperidin-1-yl)ethan-1-one

  • MDL number MFCD01700407
  • Cas Number : 93865-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74265

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74265
Availability details for
Quantity
  • Availability
  • Add to Cart