ENA321762695 Enamine

1-[(5-cyclopropyl-1,2,4-oxadiazol-3-yl)methyl]piperazine

  • MDL number MFCD14679526
  • Cas Number : 1250650-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74267
Availability details for
Quantity
  • Availability
  • Add to Cart