ENA321762697 Enamine

2-(chloromethyl)-5-cyclobutyl-1,3,4-oxadiazole

  • MDL number MFCD09051884
  • Cas Number : 1178318-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74269

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74269
Availability details for
Quantity
  • Availability
  • Add to Cart