ENA321762699 Enamine

({bicyclo[2.2.1]hept-5-en-2-yl}methyl)(propan-2-yl)amine

  • MDL number MFCD01475378
  • Cas Number : 22441-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74271
Availability details for
Quantity
  • Availability
  • Add to Cart