ENA321762701 Enamine

2-(4-chlorophenyl)-2-(piperazin-1-yl)acetamide dihydrochloride

  • MDL number MFCD18838618
  • Cas Number : 1252036-94-4
  • Molecular Weight : 326.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30342
Availability details for
Quantity
  • Availability
  • Add to Cart