ENA321762979 Enamine

1-[4-(2H-1,3-benzodioxol-5-yl)-1H-imidazol-2-yl]-3-methylbutan-1-amine dihydrochloride

  • MDL number MFCD18483295
  • Cas Number : 1311314-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74339
Availability details for
Quantity
  • Availability
  • Add to Cart