ENA321762980 Enamine

2-amino-N-(pyridin-2-yl)ethane-1-sulfonamide hydrochloride

  • MDL number MFCD01728694
  • Cas Number : 1195650-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74346
Availability details for
Quantity
  • Availability
  • Add to Cart