ENA321762984 Enamine

4-methyl-6-(thiophen-2-yl)-1,3,5-triazin-2-amine

  • MDL number MFCD18483301
  • Cas Number : 1311317-56-2
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74394
Availability details for
Quantity
  • Availability
  • Add to Cart