ENA321762988 Enamine

1-(5-chloropentyl)piperidine hydrochloride

  • MDL number MFCD18483304
  • Cas Number : 91976-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74409
Availability details for
Quantity
  • Availability
  • Add to Cart