ENA321762989 Enamine

1-(4,5-dimethoxy-2-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD18483305
  • Cas Number : 104174-56-3
  • Molecular Weight : 231.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74415
Availability details for
Quantity
  • Availability
  • Add to Cart