ENA321762993 Enamine

1-(3-bromo-4-methoxyphenyl)-2-[(3-methyl-1H-1,2,4-triazol-5-yl)sulfanyl]ethan-1-one

  • MDL number MFCD13226232
  • Cas Number : 898639-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74435

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74435
Availability details for
Quantity
  • Availability
  • Add to Cart