ENA321763001 Enamine

2-(4-aminobutyl)-2,3-dihydro-1H-isoindole-1,3-dione hydrochloride

  • MDL number MFCD18483316
  • Cas Number : 35517-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-207
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74461
Availability details for
Quantity
  • Availability
  • Add to Cart