ENA321763009 Enamine

benzyl 3-(chlorosulfonyl)piperidine-1-carboxylate

  • MDL number MFCD18483324
  • Cas Number : 1311317-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74537

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74537
Availability details for
Quantity
  • Availability
  • Add to Cart