ENA321763012 Enamine

4-[4-(1H-pyrazol-1-yl)phenyl]-1,3-thiazol-2-amine

  • MDL number MFCD18785465
  • Cas Number : 1311313-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74656
Availability details for
Quantity
  • Availability
  • Add to Cart