ENA321763016 Enamine

4-amino-1-(2,3-dihydro-1H-indol-1-yl)butan-1-one hydrochloride

  • MDL number MFCD18785466
  • Cas Number : 1311314-84-7
  • Molecular Weight : 240.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74677
Availability details for
Quantity
  • Availability
  • Add to Cart