ENA321763022 Enamine

2-amino-N-(oxan-2-ylmethyl)ethane-1-sulfonamide hydrochloride

  • MDL number MFCD18838684
  • Cas Number : 1306604-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74709

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74709
Availability details for
Quantity
  • Availability
  • Add to Cart