ENA321763024 Enamine

methyl 4-(4-aminophenoxy)butanoate hydrochloride

  • MDL number MFCD18785470
  • Cas Number : 1311314-93-8
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74728
Availability details for
Quantity
  • Availability
  • Add to Cart