ENA321763027 Enamine

tert-butyl N-[(N'-hydroxycarbamimidoyl)[2-(trifluoromethoxy)phenyl]methyl]carbamate

  • Cas Number : 1311319-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74751
Availability details for
Quantity
  • Availability
  • Add to Cart