ENA321763029 Enamine

[4-(piperidin-1-yl)oxan-4-yl]methanamine dihydrochloride

  • MDL number MFCD18483336
  • Cas Number : 1311315-87-3
  • Molecular Weight : 271.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-74757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74757
Availability details for
Quantity
  • Availability
  • Add to Cart