ENA321763030 Enamine

2-(pyridin-2-yl)-1H-1,3-benzodiazol-5-amine dihydrochloride

  • MDL number MFCD18785472
  • Cas Number : 1311316-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74758
Availability details for
Quantity
  • Availability
  • Add to Cart