ENA321763035 Enamine

tert-butyl N-[1-(2-hydroxyphenyl)propyl]carbamate

  • MDL number MFCD18785473
  • Cas Number : 1311313-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74798
Availability details for
Quantity
  • Availability
  • Add to Cart