ENA321763036 Enamine

N-(4-iodo-3-methoxyphenyl)acetamide

  • MDL number MFCD18483341
  • Cas Number : 1311316-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74803
Availability details for
Quantity
  • Availability
  • Add to Cart