ENA321763038 Enamine

6-(benzyloxy)quinoline

  • MDL number MFCD18483342
  • Cas Number : 108825-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74806

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74806
Availability details for
Quantity
  • Availability
  • Add to Cart