ENA321763039 Enamine

1-[4-(2H-1,3-benzodioxol-5-yl)-1H-imidazol-2-yl]ethan-1-amine hydrochloride

  • MDL number MFCD18483343
  • Cas Number : 1311317-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74813
Availability details for
Quantity
  • Availability
  • Add to Cart