ENA321763044 Enamine

1-[4-(2H-1,3-benzodioxol-5-yl)-1H-imidazol-2-yl]butan-1-amine dihydrochloride

  • MDL number MFCD18785477
  • Cas Number : 1311317-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74852

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74852
Availability details for
Quantity
  • Availability
  • Add to Cart