ENA321763047 Enamine

5-sulfamoylthiophene-2-carboxamide

  • MDL number MFCD18785480
  • Cas Number : 850340-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74866
Availability details for
Quantity
  • Availability
  • Add to Cart