ENA321763051 Enamine

2-[(5-chloro-2,4-dimethoxyphenyl)(2-hydroxyethyl)amino]ethan-1-ol

  • MDL number MFCD18785483
  • Cas Number : 1311315-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74886
Availability details for
Quantity
  • Availability
  • Add to Cart