ENA321763052 Enamine

ethyl 4-(chloromethyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD18785484
  • Cas Number : 54449-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74890
Availability details for
Quantity
  • Availability
  • Add to Cart