ENA321763053 Enamine

2-(3-chlorophenyl)-2-(propan-2-ylsulfanyl)ethan-1-amine hydrochloride

  • MDL number MFCD18785485
  • Cas Number : 1311315-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-74891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74891
Availability details for
Quantity
  • Availability
  • Add to Cart