ENA321763055 Enamine

methyl({[3-(pyridin-3-yl)-1,2,4-oxadiazol-5-yl]methyl})amine dihydrochloride

  • MDL number MFCD18785487
  • Cas Number : 1311315-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74896
Availability details for
Quantity
  • Availability
  • Add to Cart