ENA321763065 Enamine

2-amino-N-(4-hydroxycyclohexyl)-2-methylpropanamide hydrochloride

  • MDL number MFCD18785498
  • Cas Number : 1311316-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...252
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74953

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74953
Availability details for
Quantity
  • Availability
  • Add to Cart