ENA321763066 Enamine

2-[1-(difluoromethyl)-1H-1,3-benzodiazol-2-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD18785499
  • Cas Number : 1311316-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74978
Availability details for
Quantity
  • Availability
  • Add to Cart