ENA321763068 Enamine

N-cyclopropyl-3-(methylamino)propanamide hydrochloride

  • MDL number MFCD18785501
  • Cas Number : 1121527-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74981
Availability details for
Quantity
  • Availability
  • Add to Cart